missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, 99%, Thermo Scientific Chemicals

£32.66 - £368.84

Chemical Identifiers

CAS 106-44-5
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002376
InChI Key IWDCLRJOBJJRNH-UHFFFAOYSA-N
Synonym p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
PubChem CID 2879
ChEBI CHEBI:17847
IUPAC Name 4-methylphenol
SMILES CC1=CC=C(C=C1)O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11458690
View Documents
Thermo Scientific Alfa Aesar
100 g
£32.66
100g
Please to purchase this item. Need a web account? Register with us today!
11435643
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13531.30
250 g
£39.30
250g
Please to purchase this item. Need a web account? Register with us today!
11425643
View Documents
Thermo Scientific Alfa Aesar
A13531.0B
1000 g
£100.30
1000g
Please to purchase this item. Need a web account? Register with us today!
11445643
View Documents
Thermo Scientific Alfa Aesar
A13531.0I
5000 g
£368.84
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.

Solubility
Soluble in ethanol, diethyl ether and acetone. Slightly soluble in water.

Notes
Air and light sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents and bases.
Specifications

Chemical Identifiers

106-44-5
108.14
IWDCLRJOBJJRNH-UHFFFAOYSA-N
2879
4-methylphenol
C7H8O
MFCD00002376
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
CHEBI:17847
CC1=CC=C(C=C1)O

Specifications

106-44-5
1.034
89°C (192°F)
C7H8O
UN3455
Light sensitive
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
IWDCLRJOBJJRNH-UHFFFAOYSA-N
4-methylphenol
2879
108.14
p-Cresol
32°C to 34°C
201°C to 202°C
Phenol-like
MFCD00002376
1305151
14,2579
Soluble in ethanol,diethyl ether and acetone. Slightly soluble in water.
CC1=CC=C(C=C1)O
108.14
CHEBI:17847
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, SOLID

missing translation for 'einecsNumber' : 203-398-6

missing translation for 'rtecsNumber' : GO6475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • p-Cresol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 4-methylphenol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only