missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, 99+%, Thermo Scientific Chemicals

Product Code. 10293940
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-87-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008893
InChI Key HFPZCAJZSCWRBC-UHFFFAOYSA-N
Synonym p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
PubChem CID 7463
ChEBI CHEBI:28768
IUPAC Name 1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C

Specifications

Melting Point -68.0°C
Density 0.8540g/mL
Boiling Point 176.0°C to 178.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH(CH3)2
Refractive Index 1.4890 to 1.4910
Quantity 2.5 L
Beilstein 05, II, 322
Merck Index 15, 2760
Specific Gravity 0.854
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,chloroform,acetone,,benzene and ccl4
Formula Weight 134.22
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material p-Cymene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Cymene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H331-Toxic if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.