missing translation for 'onlineSavingsMsg'
Learn More

p-Dioxane, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

£262.00 - £1050.00

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15588544
View Documents
Avantor J.T.Baker
9231-01
500 mL Narrow Mouth Amber Glass Bottle
£262.00
500mL
Please to purchase this item. Need a web account? Register with us today!
15508554
View Documents
Avantor J.T.Baker
9231-22
1 L Aluminum Bottle
£544.00
1 L
Please to purchase this item. Need a web account? Register with us today!
15598544
View Documents
Avantor J.T.Baker
9231-03
4 L Narrow Mouth Amber Glass Bottle
£1050.00
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

123-91-1
88.106
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
CHEBI:47032
C1COCCO1
C4H8O2
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
1,4-dioxane

Specifications

123-91-1
1L = 1.03kg
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
C1COCCO1
88.106
CHEBI:47032
BAKER ANALYZED™ ACS Reagent
12.00°C
C4H8O2
RYHBNJHYFVUHQT-UHFFFAOYSA-N
1,4-dioxane
31275
88.11
p-Dioxane
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.