missing translation for 'onlineSavingsMsg'
Learn More
p-Phenylenediamine, 97%, Thermo Scientific Chemicals Product Code.: 11437627

p-Phenylenediamine, 97%, Thermo Scientific Chemicals

Product Code. 11437627
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11437627

Brand: Thermo Scientific Alfa Aesar A15680.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-50-3 | C6H8N2 | 108.144 g/mol

p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

Solubility
Soluble in alcohol, chloroform, ether and hot benzene.

Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-50-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007901
InChI Key CBCKQZAAMUWICA-UHFFFAOYSA-N
Synonym p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
PubChem CID 7814
ChEBI CHEBI:51403
IUPAC Name benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N

Specifications

Melting Point 138°C to 142°C
Boiling Point 267°C to 271°C
Flash Point 156°C (312°F)
Assay Percent Range 97%
Quantity 250 g
UN Number UN1673
Beilstein 742029
Merck Index 14,7285
Solubility Information Soluble in alcohol,chloroform,ether and hot benzene.
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material p-Phenylenediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247833 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA15680.0B,A15680.30,A15680.0I
Lot Number10245668 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberA15680.0B,A15680.30,A15680.0I
Lot Number10237828 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15680.0B,A15680.30,A15680.0I
Lot Number10146490 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15680.0B,A15680.30,A15680.0I
Lot Number5002A03R Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15680.0B,A15680.30,A15680.0I
compliance-icons compliance-icons
Product Identifier
  • p-Phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-Benzenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Phenylenediamine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.