missing translation for 'onlineSavingsMsg'
Learn More
p-Phenylenediamine, 99+%, Thermo Scientific Chemicals Product Code.: 10754163

p-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Product Code. 10754163
5 g, Glass bottle
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10754163

Brand: Thermo Scientific Acros 130570050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-50-3 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-50-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007901
InChI Key CBCKQZAAMUWICA-UHFFFAOYSA-N
Synonym p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
PubChem CID 7814
ChEBI CHEBI:51403
IUPAC Name benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N

Specifications

Melting Point 139.0°C to 141.0°C
Color Brown to Purple or Gray-White
Flash Point 156°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(NH2)2
Quantity 5 g
Beilstein 13, 61
Merck Index 15, 7397
Solubility Information Solubility in water: 47g/L (25°C). Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 108.14
Percent Purity 99+%
Physical Form Powder or Flakes
Chemical Name or Material p-Phenylenediamine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Phenylenediamine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.