Learn More
p-Phenylenediamine dihydrochloride, 99%, Thermo Scientific™
£20.49 - £210.00
Chemical Identifiers
CAS | 624-18-0 |
---|---|
Molecular Formula | C6H10Cl2N2 |
Molecular Weight (g/mol) | 181.06 |
MDL Number | MFCD00013002 |
InChI Key | IYXMNTLBLQNMLM-UHFFFAOYSA-N |
Synonym | benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64 |
PubChem CID | 12205 |
IUPAC Name | benzene-1,4-diamine;dihydrochloride |
SMILES | C1=CC(=CC=C1N)N.Cl.Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
624-18-0 | |
181.06 | |
IYXMNTLBLQNMLM-UHFFFAOYSA-N | |
12205 | |
C1=CC(=CC=C1N)N.Cl.Cl |
C6H10Cl2N2 | |
MFCD00013002 | |
benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64 | |
benzene-1,4-diamine;dihydrochloride |
Specifications
624-18-0 | |
C6H10Cl2N2 | |
UN2811 | |
14,7285 | |
IYXMNTLBLQNMLM-UHFFFAOYSA-N | |
benzene-1,4-diamine;dihydrochloride | |
12205 | |
99% |
∼265°C | |
MFCD00013002 | |
3911749 | |
benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64 | |
C1=CC(=CC=C1N)N.Cl.Cl | |
181.06 | |
181.07 | |
p-Phenylenediamine dihydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H331-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 210-834-9
missing translation for 'rtecsNumber' : ST0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- p-Phenylenediamine dihydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P361-Take off immediately all contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.