Learn More
p-Toluenesulfinic acid sodium salt, 97% (dry wt.), water <5%, Thermo Scientific Chemicals
£21.30 - £111.00
Chemical Identifiers
CAS | 824-79-3 |
---|---|
Molecular Formula | C7H7NaO2S |
Molecular Weight (g/mol) | 178.181 |
MDL Number | MFCD00149640 |
InChI Key | KFZUDNZQQCWGKF-UHFFFAOYSA-M |
Synonym | sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech |
PubChem CID | 2723791 |
IUPAC Name | sodium;4-methylbenzenesulfinate |
SMILES | CC1=CC=C(C=C1)S(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404010
|
Thermo Scientific Alfa Aesar
A11430.14 |
25 g |
£21.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400283
|
Thermo Scientific Alfa Aesar
A11430.22 |
100 g |
£33.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410283
|
Thermo Scientific Alfa Aesar
A11430.36 |
500 g |
£111.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p-Toluenesulfinic acid sodium salt is used as an intermediate for the preparation of pharmaceuticals and dispersal dyes. It acts as a curing agent of grouting material. It is also used as a discharge medium of dyes and pigments. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Toluenesulfinic acid sodium salt is used as an intermediate for the preparation of pharmaceuticals and dispersal dyes. It acts as a curing agent of grouting material. It is also used as a discharge medium of dyes and pigments. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
824-79-3 | |
178.181 | |
KFZUDNZQQCWGKF-UHFFFAOYSA-M | |
2723791 | |
CC1=CC=C(C=C1)S(=O)[O-].[Na+] |
C7H7NaO2S | |
MFCD00149640 | |
sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech | |
sodium;4-methylbenzenesulfinate |
Specifications
824-79-3 | |
C7H7NaO2S | |
4621454 | |
Slightly soluble in water. | |
CC1=CC=C(C=C1)S(=O)[O-].[Na+] | |
178.181 | |
178.19 | |
(dry wt.) |
>300°C | |
MFCD00149640 | |
sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech | |
KFZUDNZQQCWGKF-UHFFFAOYSA-M | |
sodium;4-methylbenzenesulfinate | |
2723791 | |
97% | |
p-Toluenesulfinic acid sodium salt, Water <5% |
Safety and Handling
missing translation for 'einecsNumber' : 212-538-5
missing translation for 'rtecsNumber' : XT4725000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only