Learn More
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
£38.50 - £140.00
Chemical Identifiers
CAS | 207801-20-5 |
---|---|
Molecular Formula | C7H7NaO2S |
Molecular Weight (g/mol) | 178.18 |
MDL Number | MFCD00149640 |
InChI Key | KFZUDNZQQCWGKF-UHFFFAOYSA-M |
Synonym | sodium p-toluenesulfinate hydrate, benzenesulfinic acid, 4-methyl-, sodium salt, hydrate, c7h7o2s.na.h2o, p-toluenesulfinic acid sodium salt, sodium 4-toluenesulphinate monohydrate, p-toluenesulfinic acid sodium salt hydrate, sodium 4-methylbenzenesulfinate hydrate, p-toluenesulfinic acid sodium salt dry wt. , water |
PubChem CID | 23682957 |
IUPAC Name | sodium;4-methylbenzenesulfinate;hydrate |
SMILES | [Na+].CC1=CC=C(C=C1)S([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10553111
|
Thermo Scientific Acros
139011000 |
100 g | Plastic bottle |
£38.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123240
|
Thermo Scientific Acros
139015000 |
500 g | Plastic bottle |
£140.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207801-20-5 | |
178.18 | |
KFZUDNZQQCWGKF-UHFFFAOYSA-M | |
23682957 | |
[Na+].CC1=CC=C(C=C1)S([O-])=O |
C7H7NaO2S | |
MFCD00149640 | |
sodium p-toluenesulfinate hydrate, benzenesulfinic acid, 4-methyl-, sodium salt, hydrate, c7h7o2s.na.h2o, p-toluenesulfinic acid sodium salt, sodium 4-toluenesulphinate monohydrate, p-toluenesulfinic acid sodium salt hydrate, sodium 4-methylbenzenesulfinate hydrate, p-toluenesulfinic acid sodium salt dry wt. , water | |
sodium;4-methylbenzenesulfinate;hydrate |
Specifications
207801-20-5 | |
White | |
98+% | |
CH3C6H4SO2Na·xH2O | |
11, 9 | |
10, 9360 | |
Solubility in water: soluble | |
[Na+].CC1=CC=C(C=C1)S([O-])=O | |
178.18 | |
178.18 | |
Crystalline Powder or Flakes |
>300°C | |
Authentic | |
C7H7NaO2S | |
MFCD00149640 | |
08,488 | |
sodium p-toluenesulfinate hydrate, benzenesulfinic acid, 4-methyl-, sodium salt, hydrate, c7h7o2s.na.h2o, p-toluenesulfinic acid sodium salt, sodium 4-toluenesulphinate monohydrate, p-toluenesulfinic acid sodium salt hydrate, sodium 4-methylbenzenesulfinate hydrate, p-toluenesulfinic acid sodium salt dry wt. , water | |
KFZUDNZQQCWGKF-UHFFFAOYSA-M | |
sodium;4-methylbenzenesulfinate;hydrate | |
23682957 | |
98+% | |
p-Toluenesulfinic acid,sodium salt hydrate, 98+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-538-5
missing translation for 'rtecsNumber' : XT4725000
missing translation for 'tsca' : TSCA
- p-Toluenesulfinic acid, sodium salt hydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only