Learn More
p-Toluenesulfonic Acid, 12% in Acetic Acid, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 421222500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Toluenesulfonic acid | |
12% in Acetic Acid | |
116.0°C | |
MFCD00064387 | |
11 to 13% (Alkalimetry) | |
1.0700g/mL | |
p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, tosylic acid, benzenesulfonic acid, 4-methyl, p-methylbenzenesulfonic acid | |
JOXIMZWYDAKGHI-UHFFFAOYSA-N | |
4-methylbenzenesulfonic acid | |
6101 | |
172.2 | |
Glass bottle | |
1.07 | |
41°C |
Colorless to Yellow | |
104-15-4, 64-19-7 | |
C7H8O3S | |
11, 97 | |
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507 | |
15, 9692 | |
Solubility in water: soluble. | |
CC1=CC=C(C=C1)S(=O)(=O)O | |
172.198 | |
CHEBI:27849 | |
Pure | |
1.3825 to 1.3845 | |
250 g | |
Solution |
Safety and Handling
- p-Toluenesulfonic acid, 12% in acetic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only