missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
11.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 100.0°C to 108.0°C |
Flash Point | 180°C |
Molecular Formula | C7H8O3S·H2O |
Linear Formula | CH3C6H4SO3H·H2O |
Quantity | 25 g |
Beilstein | 11,97 |
Fieser | 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507 |
Merck Index | 15,9692 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform |
Molecular Weight (g/mol) | 190.22 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- p-Toluenesulfonic acid monohydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'