Learn More
p-Toluenesulfonic acid zinc salt hydrate, Thermo Scientific Chemicals
£96.50
Chemical Identifiers
CAS | 123334-05-4 |
---|---|
Molecular Formula | C14H14O6S2Zn |
Molecular Weight (g/mol) | 407.76 |
MDL Number | MFCD00151013 |
InChI Key | YISPIDBWTUCKKH-UHFFFAOYSA-L |
Synonym | zinc p-toluenesulfonate hydrate, acmc-20eh0l, p-toluenesulfonic acid zinc salt, zinc 2+ hydrate ditosylate, p-toluenesulfonic acid zinc salt hydrate, benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:? |
PubChem CID | 16212267 |
IUPAC Name | zinc;4-methylbenzenesulfonate;hydrate |
SMILES | [Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377977
|
Thermo Scientific Alfa Aesar
020899.22 |
100 g | N/A | ||||||
Description
p-Toluenesulfonic acid zinc salt hydrate is used as a zinc benzenesulfonate salt for proteomics research, and as s a stabilizer & lubricants & activator, it's used in synthetic rubber of PE, PVC, polypropylene nylon, ABS natural rubber, butylbenzene rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123334-05-4 | |
407.76 | |
YISPIDBWTUCKKH-UHFFFAOYSA-L | |
16212267 | |
[Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O |
C14H14O6S2Zn | |
MFCD00151013 | |
zinc p-toluenesulfonate hydrate, acmc-20eh0l, p-toluenesulfonic acid zinc salt, zinc 2+ hydrate ditosylate, p-toluenesulfonic acid zinc salt hydrate, benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:? | |
zinc;4-methylbenzenesulfonate;hydrate |
Specifications
123334-05-4 | |
MFCD00151013 | |
Insoluble in water. | |
[Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O | |
407.76 | |
407.76 | |
p-Toluenesulfonic acid zinc salt hydrate |
C14H14O6S2Zn | |
zinc p-toluenesulfonate hydrate, acmc-20eh0l, p-toluenesulfonic acid zinc salt, zinc 2+ hydrate ditosylate, p-toluenesulfonic acid zinc salt hydrate, benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:? | |
YISPIDBWTUCKKH-UHFFFAOYSA-L | |
zinc;4-methylbenzenesulfonate;hydrate | |
16212267 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 236-576-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only