missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 11421928

p-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11421928
250 g
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11421928

Brand: Thermo Scientific Alfa Aesar A14547.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p-Toluenesulfonyl chloride is used as a precursor in the production of dyes and saccharin. It acts as a dehydrating agent in the conversion of urea to carbodiimide and converts alcohols into the corresponding toluenesulfonate esters. It is also employed as a flow-promoting agent for paints, an adhesive, nitrocellulose, coating material and as a plasticizer for polyamides. Furthermore, it serves as an antistatic agent, as a gloss enhancer in plastic film preparations and as a basic material of electroplating solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonyl chloride is used as a precursor in the production of dyes and saccharin. It acts as a dehydrating agent in the conversion of urea to carbodiimide and converts alcohols into the corresponding toluenesulfonate esters. It is also employed as a flow-promoting agent for paints, an adhesive, nitrocellulose, coating material and as a plasticizer for polyamides. Furthermore, it serves as an antistatic agent, as a gloss enhancer in plastic film preparations and as a basic material of electroplating solutions.

Solubility
Soluble in alcohol, benzene and ether. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
PubChem CID 7397
IUPAC Name 4-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 67°C to 70°C
Density 1.35
Boiling Point 146°C (15 mmHg)
Flash Point 128°C (262°F)
Quantity 250 g
UN Number UN3261
Beilstein 607898
Sensitivity Moisture sensitive
Merck Index 14,9534
Solubility Information Soluble in alcohol,benzene and ether; Insoluble in water.
Formula Weight 190.65
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252658 Certificate TypeCertificate of Analysis Date11/10/2024 Catalog NumberA14547.PG-100KG,A14547.30,A14547.0I,A14547.36,A14547.0B
Lot Number10249043 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog NumberA14547.PG-100KG,A14547.30,A14547.0I,A14547.36,A14547.0B
Lot Number10234516 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberA14547.PG-100KG,A14547.30,A14547.0I,A14547.36,A14547.0B
Lot Number10231289 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberA14547.PG-100KG,A14547.30,A14547.0I,A14547.36,A14547.0B
Lot Number10213047 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberA14547.PG-100KG,A14547.30,A14547.0I,A14547.36,A14547.0B
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : p-Toluene sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.