missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11431928
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g

p-Toluenesulfonyl chloride is used as a precursor in the production of dyes and saccharin. It acts as a dehydrating agent in the conversion of urea to carbodiimide and converts alcohols into the corresponding toluenesulfonate esters. It is also employed as a flow-promoting agent for paints, an adhesive, nitrocellulose, coating material and as a plasticizer for polyamides. Furthermore, it serves as an antistatic agent, as a gloss enhancer in plastic film preparations and as a basic material of electroplating solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonyl chloride is used as a precursor in the production of dyes and saccharin. It acts as a dehydrating agent in the conversion of urea to carbodiimide and converts alcohols into the corresponding toluenesulfonate esters. It is also employed as a flow-promoting agent for paints, an adhesive, nitrocellulose, coating material and as a plasticizer for polyamides. Furthermore, it serves as an antistatic agent, as a gloss enhancer in plastic film preparations and as a basic material of electroplating solutions.

Solubility
Soluble in alcohol, benzene and ether. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
PubChem CID 7397
IUPAC Name 4-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 67°C to 70°C
Density 1.35
Boiling Point 146°C (15 mmHg)
Flash Point 128°C (262°F)
Quantity 5000 g
UN Number UN3261
Beilstein 607898
Sensitivity Moisture sensitive
Merck Index 14,9534
Solubility Information Soluble in alcohol,benzene and ether; Insoluble in water.
Formula Weight 190.65
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'