missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals Product Code.: 10443883

p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals

Product Code. 10443883
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
10 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10443883

Brand: Thermo Scientific Acros 139031000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chlorideShow MoreShow Less
PubChem CID 7397
IUPAC Name 4-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 67.0°C to 69.0°C
Color White
Boiling Point 134.0°C (10.0 mmHg)
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 99% min
Packaging Glass bottle
Linear Formula CH3C6H4SO2Cl
Quantity 100 g
Beilstein 11,103
Fieser 01,1179; 03,292; 04,510; 05,676; 06,598; 08,489; 11,536; 13,286
Merck Index 15,9693
Solubility Information Solubility in water: hydrolyses. Other solubilities: hydrolysis to acid, freely soluble in alcohol, benzene and ether
Formula Weight 190.65
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469184 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number139030250,139035000,139030100,139030020,139031000
Lot NumberA0469183 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number139030250,139035000,139030100,139030020,139031000
Lot NumberA0464119 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number139030250,139030100,139035000,139030020,139031000
Lot NumberA0455369 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number139030250,139030100,139035000,139030020,139031000
Lot NumberA0402054 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number139030250,139030100,139035000,139030020,139031000
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.