missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals

Product Code. 10775981
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
10 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
PubChem CID 7397
IUPAC Name 4-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 67.0°C to 69.0°C
Color White
Boiling Point 134.0°C (10.0 mmHg)
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 99% min
Packaging Glass bottle
Linear Formula CH3C6H4SO2Cl
Quantity 2 kg
Beilstein 11,103
Fieser 01,1179; 03,292; 04,510; 05,676; 06,598; 08,489; 11,536; 13,286
Merck Index 15,9693
Solubility Information Solubility in water: hydrolyses. Other solubilities: hydrolysis to acid, freely soluble in alcohol, benzene and ether
Formula Weight 190.65
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'