missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonyl fluoride, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Melting Point | 40.0°C to 43.0°C |
Boiling Point | 112.0°C (16.0 mmHg) |
Flash Point | 106°C |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | CH3C6H4SO2F |
Quantity | 5 g |
Beilstein | 11, II, 54 |
Solubility Information | Solubility in water: hydrolysis. |
Formula Weight | 174.19 |
Show More |
Safety and Handling


Product Identifier
- p-Toluenesulfonyl fluoride
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
p-Toluenesulfonyl fluoride, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction