missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonyl fluoride, 98%, Thermo Scientific Chemicals Product Code.: 11398665

p-Toluenesulfonyl fluoride, 98%, Thermo Scientific Chemicals

Product Code. 11398665
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11398665

Brand: Thermo Scientific Alfa Aesar L12204.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p-Toluenesulfonyl fluoride is used for sulfonylation of organometallic reagents as a simple, one-step synthesis of sulfones. It is useful in the treatment of Alzheimer?s disease. It is used to study the DNS polymerase from spinach leaf extracts. It was used in isolation and separation of Pseudomonas aeruginosa membranes by density sucrose gradient centrifugation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-16-3
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00007421
InChI Key IZZYABADQVQHLC-UHFFFAOYSA-N
Synonym p-toluenesulfonyl fluoride, tosyl fluoride, 4-toluenesulfonyl fluoride, benzenesulfonyl fluoride, 4-methyl, 4-tolylsulfonyl fluoride, toluene-4-sulfonyl fluoride, p-methylbenzenesulfonyl fluoride, p-toluenesulphonyl fluoride, toluene-p-sulphonyl fluoride, p-methylphenylsulfonyl fluoride
PubChem CID 9965
IUPAC Name 4-methylbenzenesulfonyl fluoride
SMILES CC1=CC=C(C=C1)S(=O)(=O)F

Specifications

Melting Point 40°C to 41°C
Boiling Point 112°C (16 mmHg)
Flash Point 105°C (221°F)
Quantity 25 g
UN Number UN3261
Beilstein 2208423
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 174.19
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonyl fluoride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.