missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonyl isocyanate, 96%, Thermo Scientific Chemicals Product Code.: 10265520

p-Toluenesulfonyl isocyanate, 96%, Thermo Scientific Chemicals

Product Code. 10265520
25 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10265520

Brand: Thermo Scientific Acros 183070250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4083-64-1
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00002030
InChI Key VLJQDHDVZJXNQL-UHFFFAOYSA-N
Synonym tosyl isocyanate, p-toluenesulfonyl isocyanate, 4-methylbenzenesulfonyl isocyanate, p-toluenesulphonyl isocyanate, benzenesulfonyl isocyanate, 4-methyl, tosylisocyanate, p-tolylsulfonyl isocyanate, unii-h9004fjx6v, p-toluenesulfonic acid, anhydride with isocyanic acid, p-toluene sulfonyl isocyanate
PubChem CID 77703
IUPAC Name 4-methyl-N-(oxomethylidene)benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Melting Point -2°C
Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 144°C (10.0 mmHg)
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SO2NCO
Refractive Index 1.5330 to 1.535
Quantity 25 mL
Fieser 07,377
Specific Gravity 1.29
Solubility Information Solubility in water: reacts
Formula Weight 197.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material p-Toluenesulfonyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465379 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number183071000,183070250,183075000
Lot NumberA0463568 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number183071000,183070250,183075000
Lot NumberA0460191 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number183071000,183070250,183075000
Lot NumberA0458068 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number183071000,183070250,183075000
Lot NumberA0453606 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number183071000,183070250,183075000
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonyl isocyanate, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.