missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl semicarbazide, 95%, Thermo Scientific Chemicals

Product Code. 15438538
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10396-10-8
Molecular Formula C8H11N3O3S
Molecular Weight (g/mol) 229.254
MDL Number MFCD00072243
InChI Key VRFNYSYURHAPFL-UHFFFAOYSA-N
Synonym p-toluenesulfonylsemicarbazide, 1-tosylsemicarbazide, p-toluenesulfonyl semicarbazide, benzenesulfonic acid, 4-methyl-, 2-aminocarbonyl hydrazide, 1-p-methylphenylsulfonyl semicarbazide, 4-methylbenzenesulfonamidourea, p-toluenesulfohyl semicarbazide, 4-methylbenzenesulfonamido urea, 4-toluenesulfonyl semicarbazide, ra ptss
PubChem CID 82602
IUPAC Name [(4-methylphenyl)sulfonylamino]urea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NNC(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 229.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material p-Toluenesulfonyl semicarbazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'