missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 117°C to 119°C |
Quantity | 5 g |
Beilstein | 2331311 |
Solubility Information | Insoluble in water. |
Formula Weight | 214.24 |
Percent Purity | 98% |
Chemical Name or Material | p-Toluenesulfonylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'