missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonylacetonitrile, 98+%, Thermo Scientific Chemicals Product Code.: 11455273

p-Toluenesulfonylacetonitrile, 98+%, Thermo Scientific Chemicals

Product Code. 11455273
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11455273

Brand: Thermo Scientific Alfa Aesar A12857.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5697-44-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00039487
InChI Key BBNNLJMGPASZPD-UHFFFAOYSA-N
Synonym p-toluenesulfonylacetonitrile, 2-tosylacetonitrile, tosylacetonitrile, p-tolylsulfonylacetonitrile, 4-methylphenyl sulfonyl acetonitrile, 4-toluenesulfonylacetonitrile, 4-methylphenyl sulfonylacetonitrile, 4-toluenesulphonylacetonitrile, p-cyanomethylsulfonyl toluene, tosylmethyl isocyanideShow MoreShow Less
PubChem CID 79776
IUPAC Name 2-(4-methylphenyl)sulfonylacetonitrile
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC#N

Specifications

Melting Point 147°C to 152°C
Quantity 100 g
UN Number UN3439
Beilstein 1842131
Formula Weight 195.24
Percent Purity ≥98%
Chemical Name or Material p-Toluenesulfonylacetonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10189002 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog NumberA12857.22,A12857.06,A12857.14
Lot NumberJ2058A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12857.22,A12857.06,A12857.14
Lot Number10189002 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12857.22,A12857.06,A12857.14
Lot Number28790 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12857.22,A12857.06,A12857.14
Lot NumberG6094B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12857.22,A12857.06,A12857.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonylacetonitrile, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.