missing translation for 'onlineSavingsMsg'
Learn More
p-Toluic acid, 98%, Thermo Scientific Chemicals Product Code.: 10366590

p-Toluic acid, 98%, Thermo Scientific Chemicals

Product Code. 10366590
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10366590

Brand: Thermo Scientific Acros 139061000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-94-5
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002565
InChI Key LPNBBFKOUUSUDB-UHFFFAOYSA-N
Synonym p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
PubChem CID 7470
ChEBI CHEBI:36635
IUPAC Name 4-methylbenzoic acid
SMILES CC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 179.0°C to 182.0°C
Color Cream-Yellow to White
Boiling Point 274.0°C to 275.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula CH3C6H4CO2H
Quantity 100 g
Beilstein 09,483
Merck Index 14,9535
Solubility Information Solubility in water: 11.6g/L (98°C). Other solubilities: soluble in alcohol and ether
Formula Weight 136.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material p-Toluic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.