missing translation for 'onlineSavingsMsg'
Learn More

p-Toluic hydrazide, 98%, Thermo Scientific Chemicals

Product Code. 11474334
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3619-22-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007607
InChI Key MFFVZXOPRXMVET-UHFFFAOYSA-N
Synonym p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide
PubChem CID 77174
IUPAC Name 4-methylbenzohydrazide
SMILES CC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 115°C to 118°C
Quantity 100 g
Beilstein 607602
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material p-Toluic hydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'