missing translation for 'onlineSavingsMsg'
Learn More
p-Toluidine, 99%, flakes, ACROS Organics™ Product Code.: 10591571

p-Toluidine, 99%, flakes, ACROS Organics™

Product Code. 10591571
1kg; Glass bottle
Click to view available options
1kg; Glass bottle
This item is not returnable. View return policy

Product Code. 10591571

Brand: Thermo Scientific Acros 139090010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 106-49-0
Melting Point 41°C to 46°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H9N
Linear Formula CH3C6H4NH2
MDL Number MFCD00007906
Quantity 1 kg
Beilstein 12, 880
Merck Index 15, 9696
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)N
IUPAC Name 4-methylaniline
Molecular Weight (g/mol) 107.156
PubChem CID 7813
ChEBI CHEBI:37825
Formula Weight 107.16
Percent Purity ≥98.5%
Physical Form Flakes
Chemical Name or Material p-Toluidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluidine, 99%, flakes, ACROS Organics™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.