missing translation for 'onlineSavingsMsg'
Learn More

p-Toluidine, 99+%

Product Code. 11454684
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11454684

Brand: Thermo Scientific Alfa Aesar A13698.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 106-49-0 | C7H9N | 107.156 g/mol

p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

Solubility
Soluble in ethanol, pyridine, diethyl ether, acetone, carbon tetrachloride, methanol, carbon disulfide, oils and dilute acids.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-49-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00007906
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
PubChem CID 7813
ChEBI CHEBI:37825
IUPAC Name 4-methylaniline
SMILES CC1=CC=C(C=C1)N

Specifications

Melting Point 42°C to 46°C
Density 1.05
Boiling Point 199°C to 202°C
Flash Point 88°C (190°F)
Quantity 1000 g
UN Number UN3451
Beilstein 471281
Sensitivity Air and light sensitive
Merck Index 14,9536
Solubility Information Soluble in ethanol,pyridine,diethyl ether,acetone,carbon tetrachloride,methanol,carbon disulfide,oils and dilute acids.
Formula Weight 107.16
Percent Purity ≥99%
Chemical Name or Material p-Toluidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.