missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetyl chloride, ≥97%, Thermo Scientific™

£67.70 - £128.00

Chemical Identifiers

CAS 35675-44-6
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00018813
InChI Key QDZAWVLWIMOXJT-UHFFFAOYSA-N
Synonym p-tolylacetyl chloride, 2-4-methylphenyl acetyl chloride, p-tolyl-acetyl chloride, 4-methyl-phenyl-acetyl chloride, 4-methylphenylacetylchloride, p-methylphenylacetyl chloride, 2-4-tolyl acetyl chloride, 4-methylphenylacetyl chloride, 4-methylphenyl acetyl chloride, 4-methylphenylacetic acid chloride
PubChem CID 2760634
IUPAC Name 2-(4-methylphenyl)acetyl chloride
SMILES CC1=CC=C(CC(Cl)=O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427857
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H37997.MD
250 mg
£67.70
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15417857
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H37997.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

35675-44-6
168.62
QDZAWVLWIMOXJT-UHFFFAOYSA-N
2760634
CC1=CC=C(CC(Cl)=O)C=C1
C9H9ClO
MFCD00018813
p-tolylacetyl chloride, 2-4-methylphenyl acetyl chloride, p-tolyl-acetyl chloride, 4-methyl-phenyl-acetyl chloride, 4-methylphenylacetylchloride, p-methylphenylacetyl chloride, 2-4-tolyl acetyl chloride, 4-methylphenylacetyl chloride, 4-methylphenyl acetyl chloride, 4-methylphenylacetic acid chloride
2-(4-methylphenyl)acetyl chloride

Specifications

35675-44-6
115°C (7.5 mmHg)
MFCD00018813
Moisture sensitive
QDZAWVLWIMOXJT-UHFFFAOYSA-N
2-(4-methylphenyl)acetyl chloride
2760634
≥97%
p-Tolylacetyl chloride
Pink
C9H9ClO
UN3265
p-tolylacetyl chloride, 2-4-methylphenyl acetyl chloride, p-tolyl-acetyl chloride, 4-methyl-phenyl-acetyl chloride, 4-methylphenylacetylchloride, p-methylphenylacetyl chloride, 2-4-tolyl acetyl chloride, 4-methylphenylacetyl chloride, 4-methylphenyl acetyl chloride, 4-methylphenylacetic acid chloride
CC1=CC=C(CC(Cl)=O)C=C1
168.62
168.62
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications