Learn More
p-Tolylmagnesium chloride, 2M (31 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals
£173.71 - £1356.61
Chemical Identifiers
CAS | 696-61-7, 109-99-9 |
---|---|
Molecular Formula | C7H7ClMg |
Molecular Weight (g/mol) | 150.888 |
MDL Number | MFCD00672097 |
InChI Key | BLMRDOZWELZHDX-UHFFFAOYSA-M |
Synonym | p-tolylmagnesium chloride, magnesium, chloro 4-methylphenyl, 4-tolylmagnesium chloride, 4-tolyl-magnesium chloride, 4-methylphenylmagnesium chloride, ybpfqofspmwgka-uhfffaoysa-m, p-tolylmagnesium chloride solution, 1.0 m in thf |
PubChem CID | 11829629 |
IUPAC Name | magnesium;methylbenzene;chloride |
SMILES | CC1=CC=[C-]C=C1.[Mg+2].[Cl-] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
696-61-7, 109-99-9 | |
150.888 | |
BLMRDOZWELZHDX-UHFFFAOYSA-M | |
11829629 | |
CC1=CC=[C-]C=C1.[Mg+2].[Cl-] |
C7H7ClMg | |
MFCD00672097 | |
p-tolylmagnesium chloride, magnesium, chloro 4-methylphenyl, 4-tolylmagnesium chloride, 4-tolyl-magnesium chloride, 4-methylphenylmagnesium chloride, ybpfqofspmwgka-uhfffaoysa-m, p-tolylmagnesium chloride solution, 1.0 m in thf | |
magnesium;methylbenzene;chloride |
Specifications
Undesignated | |
C7H7ClMg | |
MFCD00672097 | |
1.0000g/mL | |
BLMRDOZWELZHDX-UHFFFAOYSA-M | |
magnesium;methylbenzene;chloride | |
11829629 | |
1 | |
Solution |
696-61-7, 109-99-9 | |
CH3C6H4MgCl | |
2 to 2.2M | |
p-tolylmagnesium chloride, magnesium, chloro 4-methylphenyl, 4-tolylmagnesium chloride, 4-tolyl-magnesium chloride, 4-methylphenylmagnesium chloride, ybpfqofspmwgka-uhfffaoysa-m, p-tolylmagnesium chloride solution, 1.0 m in thf | |
CC1=CC=[C-]C=C1.[Mg+2].[Cl-] | |
150.888 | |
150.89 | |
−17°C | |
p-Tolylmagnesium chloride, 2M solution in THF |
Safety and Handling
GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- p-Tolylmagnesium chloride, 2M (31 wt.%) solution in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only