missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyltrichlorosilane, 97%, Thermo Scientific Chemicals

£22.80 - £69.65

Chemical Identifiers

CAS 701-35-9
Molecular Formula C7H7Cl3Si
Molecular Weight (g/mol) 225.568
MDL Number MFCD00045131
InChI Key WOMUGKOOLXQCTQ-UHFFFAOYSA-N
Synonym p-tolyltrichlorosilane, tolyltrichlorosilane, trichloro-p-tolylsilane, silane, trichloro 4-methylphenyl, trichloro 4-methylphenyl silane, trichloro p-tolyl silane, silane, trichloro-p-tolyl, benzene, 1-methyl-4-trichlorosilyl, 4-tolyltrichlorosilane, p-tolylsilyl trichloride
PubChem CID 69697
IUPAC Name trichloro-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430337
View Documents
Thermo Scientific Alfa Aesar
B21728.06
5 g
£22.80
5g
Please to purchase this item. Need a web account? Register with us today!
11440337
View Documents
Thermo Scientific Alfa Aesar
B21728.14
25 g
£69.65
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

701-35-9
225.568
WOMUGKOOLXQCTQ-UHFFFAOYSA-N
69697
CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
C7H7Cl3Si
MFCD00045131
p-tolyltrichlorosilane, tolyltrichlorosilane, trichloro-p-tolylsilane, silane, trichloro 4-methylphenyl, trichloro 4-methylphenyl silane, trichloro p-tolyl silane, silane, trichloro-p-tolyl, benzene, 1-methyl-4-trichlorosilyl, 4-tolyltrichlorosilane, p-tolylsilyl trichloride
trichloro-(4-methylphenyl)silane

Specifications

218°C to 220°C
C7H7Cl3Si
UN2987
98°C (208°F)
Reacts with water.
CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
225.568
225.58
Irritating
Moisture sensitive
p-Tolyltrichlorosilane
701-35-9
MFCD00045131
1866557
p-tolyltrichlorosilane, tolyltrichlorosilane, trichloro-p-tolylsilane, silane, trichloro 4-methylphenyl, trichloro 4-methylphenyl silane, trichloro p-tolyl silane, silane, trichloro-p-tolyl, benzene, 1-methyl-4-trichlorosilyl, 4-tolyltrichlorosilane, p-tolylsilyl trichloride
WOMUGKOOLXQCTQ-UHFFFAOYSA-N
trichloro-(4-methylphenyl)silane
69697
97%
1.524
1.28
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 211-854-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only