missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene, 99%, for HPLC, Thermo Scientific Chemicals

Product Code. 11974441
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 13.0°C
Absorbance 0.01 max. at 380nm, 0.01 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 320nm, 1 max. at 294nm
Density 0.8660g/mL
Boiling Point 138.0°C
Quantity 2.5 L
Flash Point 25°C
Packaging Glass Bottle
Refractive Index 1.4940 to 1.4960
Assay Percent Range 99%
Specific Gravity 0.866
Linear Formula C6H4(CH3)2
Viscosity 0.648 mPa.s (20°C)
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Percent Purity 99%
Grade HPLC
Chemical Name or Material p-Xylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.