missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene, 99%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Tamaño de la unidad:
1 L
10 L
2.5 L
500mL
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Melting Point | 13.0°C |
| Color | Colorless to Yellow |
| Density | 0.8660g/mL |
| Boiling Point | 138.0°C |
| Quantity | 500 mL |
| Flash Point | 25°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.4940 to 1.4960 |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- p-Xylene
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido