missing translation for 'onlineSavingsMsg'
Learn More
p-Xylene, 99%, for spectroscopy Product Code.: 10056000

p-Xylene, 99%, for spectroscopy

Product Code. 10056000
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10056000

Brand: Thermo Scientific Acros 422670010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 106-42-3 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For fluorescence/luminescence spectroscopy, For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 138.0°C
Melting Point 13.0°C
CAS 106-42-3
Physical Form Liquid
Density 0.8660g/mL
Quantity 1 L
Flash Point 25°C
Assay Percent Range 99%
Infrared Spectrum Authentic
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
MDL Number MFCD00008556
Beilstein 05, 382
Merck Index 15, 10278
Packaging Glass Bottle
Synonym 1, 4-Dimethylbenzene
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
SMILES CC1=CC=C(C)C=C1
IUPAC Name 1,4-xylene
Molecular Weight (g/mol) 106.17
PubChem CID 7809
ChEBI CHEBI:27417
Formula Weight 106.17
Percent Purity 99%
Grade Spectroscopy
Specific Gravity 0.866
Chemical Name or Material p-Xylene, scintillation grade, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Xylene, 99%, for spectroscopy >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.