missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
Solubility
Miscible with alcohol, ether, acetone, benzene and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
Solubility
Miscible with alcohol, ether, acetone, benzene and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 12°C to 13°C |
Density | 0.866 |
Boiling Point | 137°C to 138°C |
Quantity | 100 mL |
Flash Point | 27°C (80°F) |
Odor | Aromatic |
Refractive Index | 1.495 |
UN Number | UN1307 |
Beilstein | 1901563 |
Merck Index | 14,10081 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'