missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylenebis(triphenylphosphonium chloride), 97+%, Thermo Scientific™

Product Code. 11354564
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1519-47-7
Molecular Formula C44H38Cl2P2
Molecular Weight (g/mol) 699.64
MDL Number MFCD00031710
InChI Key IUNJPQCPINLADS-UHFFFAOYSA-L
Synonym p-xylylenebis triphenylphosphonium chloride, 1,4-phenylenebis methylene bis triphenylphosphonium chloride, phosphonium, p-xylylenebis triphenyl-, dichloride, p-phenylenebis methylene bis triphenylphosphonium dichloride, phosphonium, p-phenylenedimethylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, 1,4-phenylenebis methylene bis triphenylphosphonium dichloride, xylylenebistriphenylphosphoniumchloride, p-xylylenebis triphenylphosphonium chloride 5g
PubChem CID 168940
IUPAC Name triphenyl-[[4-(triphenylphosphaniumylmethyl)phenyl]methyl]phosphanium;dichloride
SMILES [Cl-].[Cl-].C(C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300°C
Quantity 5 g
UN Number UN3464
Beilstein 3876684
Sensitivity Hygroscopic
Formula Weight 699.66
Percent Purity ≥97%
Chemical Name or Material p-Xylylenebis(triphenylphosphonium chloride)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'