missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical
104.12 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
HDPE plastic bottle
Description
Rready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
CAS | 7761-88-8 |
Molecular Formula | AgNO3 |
MDL Number | MFCD00003414 |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
SMILES | [Ag+].[O-][N+]([O-])=O |
IUPAC Name | silver;nitrate |
Molecular Weight (g/mol) | 169.87 |
PubChem CID | 24470 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Silver nitrate, 0.1M solution
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : Silver nitrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'