Learn More
Invitrogen™ CyQUANT™ MTT Cell Viability Assay
MTT Cell Proliferation Assay Kit simplifies the task of counting cells
Brand: Invitrogen™ V13154
289.49 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.15000kg
Description
MTT is an established reagent that is widely used to determine cell number in a variety of applications. Live cells reduce MTT to a strongly pigmented formazan product. After solubilization, the absorbance of the formazan can be measured with a microplate absorbance reader – a fluorometer is not needed.
- This kit provides complete package for performing a cell-counting assay that can detect from 2000 to 250,000 cells, depending on the cell type and conditions
- Conveniently packaged reagents reduce waste and detailed protocol conserves valuable time
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Proliferation, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, High-Content Screening (HCS), Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room temperature. The kit provides a sufficient amount of material for approximately1000 assays.
Specifications
Store at 2–8°C and protect from light. | |
Viability Assay | |
Cell Viability Assay | |
96-well plate | |
Room Temperature |
Colorimetric | |
Microplate Reader | |
Other Label(s) or Dye(s) | |
CyQUANT™ |
For Research Use Only. Not for use in diagnostic procedures.