Learn More
Thermo Scientific™ Micro BCA™ Protein Assay Kit
Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).
Brand: Thermo Scientific™ 23235
352.70 GBP valid until 2024-12-31
Use promo code "21989" to get your promotional price.
Code : PN
Additional Details : CAS Number : 7758-99-8 Weight : 1.81440kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).
The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples. Mixing together the three Micro BCA Reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0.5 to 20μg/mL protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.Highlights:
Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)
High linearity – linear working range for BSA equals 0.5 to 20μg/mL
Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)
Compatible – unaffected by typical concentrations of most ionic and nonionic detergents
Convenient – microplate and cuvette protocols provided with the instructions
Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours
Includes:
Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)
Specifications
Sufficient For: 480 tube assays or 3200 microplate assays • Micro BCA Reagent A (MA), 240 mL • Micro BCA Reagent B (MB), 240 mL • Micro BCA Reagent C (MC), 12 mL • Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL Store at room temperature. |
|
BCA Assay | |
Micro BCA™, Pierce™ | |
Not Target-Specific | |
Solution-based Detection, Absorbance | |
500 mL |
Pierce Micro BCA Protein Assay Kit | |
Colorimetric | |
Protein Quantitation Assay | |
480 Tube Assays or 3200 Microplate Assays | |
Spectrophotometer, Microplate Reader |
Safety and Handling
- SET MICRO BCA PROTEIN ASSAY KIT, 500ML KIT
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
For Research Use Only. Not for use in diagnostic procedures.