Learn More
Electrode Storage Solution, Fisher BioReagents™
For use in storage of pH electrodes
Brand: Fisher Bioreagents BP2418-1
44.79 GBP valid until 2024-12-31
Use promo code "21986" to get your promotional price.
Code : 32
Additional Details : CAS Number : 877-24-7 Weight : 0.51000kg
Description
For use in storage of pH electrodes
Electrode Storage Solution is a safe and convenient method to maintain sensitivity by removing buildup from electrodes.Specifications
Electrode Storage Solution | |
0°C | |
dihydrogen oxide, dihydrogen monoxide | |
Poly Bottle | |
14mmHg at 20°C | |
Liquid |
Undesignated | |
1g/cm³ | |
Pass Test | |
1.57 to 1.73% | |
1 L |