missing translation for 'onlineSavingsMsg'
Learn More

SI-Analytics™ TitroLine™ 7500 KF trace M2 Titrator

Increase precision with high visibility full color display and standard methods for different applications. SI Analytics™ TitroLine™ 7500 KF trace M2 Titrator possesses general titration feautres but offers improved performace.

Brand:  SI-Analytics™ 285220870


Code : NEW

Additional Details : Weight : 2.30000kg

Product Code. 13596600

  • £6190.00 / Each
Estimated Shipment: 08-10-2024
to see stock.
Add to basket

Description

Description

  • Full color display can be easily viewed from a distance and extreme angles
  • Store results via USB port (PDF - and CSV - format)
  • Includes standard methods for different applications (titer, blank value, 1 or 2 component reagent)
  • Titration stand manages the addition of solvent and titrated sample extraction
  • Accuracy: 0.3% at 1mg water
  • Includes: Coulometric KF-titrator, scope of supply: titrator unit, generator electrode TZ 1752 without diaphragm, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1754, micro-dual platinum electrode KF 11
Specifications

Specifications

RS232
Full color display
TitriSoft 3.0 PC Software
TitroLine 7500 KF trace M2
15.3 cm
1ppm – 5 % (10 μg to 100 mg)
KF coulometry, bromine number
Coulometric KF-titrator, scope of supply: titrator unit, generator electrode without diaphragm, titration stand with integrated stirrer and pump, titration vessel, micro-dual platinum electrode
10°C to 40°C
29.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers