Learn More
Invitrogen™ PureLink™ Microbiome DNA Purification Kit
The Invitrogen™ PureLink™ Microbiome DNA Purification Kit uses proven PureLink™ spin column technology for robust yields of purified DNA ready for downstream PCR, sequencing or other applications.
Brand: Invitrogen™ A29790
163.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00000kg
Description
The Invitrogen™ PureLink™ Microbiome DNA Purification Kit uses proven PureLink™ spin column technology for robust yields of purified DNA ready for downstream PCR, sequencing or other applications. The highly efficient triple lysis approach, fast removal of inhibitors, and versatility makes PureLink the ultimate kit for microbiome research projects as well as programs aimed at rapid detection of pathogenic bacteria in various samples.
- Efficient lysis of all microorganisms (including durable species with thicker and more complex cell walls) by a combination of heat, chemical, and mechanical disruption with specialized beads
- Elimination of inhibitory compounds by precipitation using a novel cleanup buffer
- Streamlined protocols for numerous biological samples
- Recovery of highly pure DNA compatible with common downstream applications such as qPCR and next-generation sequencing
Certifications
Certificates of Analysis for each lot
Services
Product performance guaranteed for 1 year from date of sale
Specifications
Spin Column | |
Silica Spin Column | |
Microbial Culture, Saliva, Soil, Stool, Host DNA from Stool, Swabs (Buccal, Vaginal, Skin, Rectal, or Environmental), Cells, Food and Environmental | |
Microbial and host DNA | |
Not High-throughput Compatible (Manual) | |
Room Temperature | |
≤25 μg |
• 40 mL S1 Lysis Buffer; room temperature • 5 mL S2 Lysis Enhancer; room temperature • 12.5 mL S3 Cleanup Buffer; room temperature • 45 mL S4 Binding Buffer; room temperature • 13 mL S5 Wash Buffer (concentrated); room temperature • 5 mL S6 Elution Buffer; room temperature • 50 PureLink Columns with Collection Tubes; room temperature • 100 PureLink Collection Tubes; room temperature • 50 Bead Tubes; room temperature |
|
Mini | |
50 to 200 μL | |
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization | |
50 Preps | |
Up to 55 min. (Including sample homogenization) | |
Buccal/Vaginal/Skin/Rectal/Environmental Swab: 1 swab Microbial Culture: ≤5 mL Saliva: ≤2 mL Soil: ≤2 g Stool: ≤2 g Urine: ≤10 mL |
For research use only. Not for use in diagnostic procedures.