Learn More
Bel-Art™ SP Scienceware™ Waste Disposal Can with Lift-up Cover
Bright, red safety disposal container suitable for biological/biohazard materials, broken glass, recyclables or other waste
Brand: Bel-Art™ F13197-0000
72.66 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.50000kg
Alert:
If using the container for biohazardous waste, then a biohazard label is required (not supplied).
Description
- Sized for benchtop or floor placement
- One piece hinged lid protects users from contact with contents; helps retain odors and contain airborne contaminants
- Molded of polypropylene for long service life
- Puncture-resistant; autoclavable at 121°C (250°F)
- Fits 19 × 23 in. autoclave bags (sold separately)
- Secured with an internal stainless steel wire bar
Specifications
Hinged Top | |
Red | |
Yes | |
22.5 cm | |
Autoclavable | |
12 in. | |
11.5 in. | |
Safety Container |
5 gal. | |
9 x 12 x 11.5 in. (22.5 x 30.5 x 29 cm) | |
29 cm | |
30.5 cm | |
Biological or biohazard materials, broken glass, recyclables or other waste | |
9 in. | |
Polypropylene |