missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile 50%, Water 47.5 % and Trifluoroacetic acid 2.5%, For mass spectrometry, Honeywell Riedel-de Haën™
76.84 GBP valid until 2024-12-31
Use promo code "21961" to get your promotional price.
Use promo code "21961" to get your promotional price.
Click to view available options
Quantity:
250 mL
Packaging:
Glass Bottle
Description
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Flash Point | <10°C |
Quantity | 250 mL |
CAS | 75-05-8, 7732-18-5, 76-05-1 |
CAS Max % | 50.0000%, 47.5000% |
Packaging | Glass Bottle |
Molecular Formula | C2H3N |
MDL Number | MFCD00001878 |
UN Number | UN1648 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Acetonitrile 50%, Water 47.5 % and TFA 2.5%
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : acetonitrile, trifluoroacetic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'