Learn More
HALYARD™ PUREZERO™ Pouch Face Mask with Headband
GREENER_CHOICE
A three-layer mask that provides high particle, bacterial, viral filtration efficiency, and expanded breathing chamber. The mask consists of SO SOFT biocomponent technology inner lining for comfort and low linting neckguard for increased protection.
Brand:  HALYARD™ CLN3941BBG
173.39 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Product Code. 17857980
  • £185.00 / Pack of 300
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to meet the stringent requirements of cleanroom environments
  • Manage user comfort and protection with SO SOFT inner lining, a foam nose band for comfort and fog reduction
  • Manage product contamination by providing high particle, bacterial, and viral filtration efficiency
Specifications
Specifications
Cleanroom Face Mask
Headband
One Size
Pouch
Store in a clean, dry environment. Avoid direct sunlight or other sources of heat.
BFE > 98%, PFE > 98%,VFE 97%
Yes
50/Bag, 10 Bags/Cs.
10 in.
White
Polyester, Polyethylene
Non-sterile
General Product Safety Directive 2001/95/EC, ISO5/6/7/8
Single-use
Cleanrooms
CLN3941
PE Bag
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
HALYARD™ PUREZERO™ Pouch Face Mask with Headband > with Headband