Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles
Save time and minimize contamination with these radiation-sterilized filter storage bottles. Excellent as single-use storage containers.
£108.00 - £190.92
Specifications
Material Virgon Clear Polystyrene Bottle, White Pigmented Virgin HDPE Closure
Sterility Sterile
Model Wide
Mouth Wide
Fit for Purpose Sterile
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10074470
View Documents Promotion Details
Thermo Scientific™
455-0500
500 mL
£108.00
Pack of 12
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10278370
View Documents Promotion Details
Thermo Scientific™
455-0150
150 mL
£190.92
Pack of 24
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10636591
View Documents Promotion Details
Thermo Scientific™
455-0250
250 mL
£163.06
Pack of 24
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10546361
View Documents Promotion Details
Thermo Scientific™
455-1000
1 L
£163.06
Pack of 12
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
Save time and minimize contamination with gamma-sterilized Thermo Scientific™ Nalgene™ Rapid-Flow™ Filter Storage Bottles. Use with Nalgene Bottle-Top filters or as replacement/extra receptacles for filter units. Excellent as single-use storage containers.
  • Headspace allows for extra serum or other additives to be added to the filtrate
  • Leakproof† screw caps to eliminate pH shift in bottle for longer life of stored media
  • Sterilized by gamma radiation and individually bagged for a 5-year sterile shelf life
  • Non-pyrogenic and non-cytotoxic
  • Available with Fit for Purpose Sterile documentation
Specifications
Specifications
Virgon Clear Polystyrene Bottle, White Pigmented Virgin HDPE Closure
Wide
Sterile
Sterile
Wide
Videos
SDS
Documents
Documents
Product Certifications
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.