missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated microplates for diverse applications
£180.00 - £540.00
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Product Code | Brand | Color | Volume (Metric) Well | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) Well | Price | Quantity & Availability | ||||
10236333
|
BRAND™
781622 |
Black | 0.1 mL |
£209.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
10678463
|
BRAND™
781620 |
Clear | 0.1 mL |
£193.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
10582474
|
BRAND™
781621 |
White | 0.1 mL |
£180.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
10752304
|
BRAND™
781626 |
White, Clear Bottom | 0.12 mL |
£540.00
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10072103
|
BRAND™
781627 |
Black, Clear Bottom | 0.12 mL |
£540.00
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
Description
- Non-treated PS surface
- Medium binding, non-sterile
- Standard plate for most applications
- Free from endotoxins, DNase, DNA, RNase; noncytoxic
Specifications
Standard | |
16 x 24 | |
384 | |
Non-treated | |
For homogenous assays, screenings, storage |
Without Lid | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |