missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Save time and money with these Concentrator Kits. Everything you need for quick drying and concentrating of small sample volumes
£7520.66
Specifications
Certifications | UL Listed to US and Canadian requirements. CE marked |
---|---|
Holds | Accommodates 36 x 1.5 mL Tubes, 72 x 0.5 mL Tubes and 2 Microwell Plates |
Includes | RD36 rotor (36x 1.5-2.0 ml tubes) |
Max. Vacuum | <10 Torr (13 mbar, 1.3k Pa) |
Shipping Weight (English) | 105 lb. |
Product Code | Brand | Description | Electrical Requirements | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Electrical Requirements | Price | Quantity & Availability | ||||
15819206
|
Thermo Scientific™
DNA130-230 |
SpeedVac DNA130 Vacuum Concentrator System | 230 V 50 Hz |
£7520.66
Each |
In Stock
Log in to see stock. |
||||
Description
Designed for fast, efficient concentration of DNA and RNA applications, the Thermo Scientific™ Savant SpeedVac™ DNA130 is a compact, integrated vacuum concentrator. SpeedVac™ DNA130 vacuum concentrator will support sample preparations with ease and efficiency.
- Compact, easy to use design: smallest integrated benchtop unit saves lab bench space.
- Corrosion-resistant, PTFE coated chamber offers years of service
- Built-in, oil free vacuum pump offers maintenance-free operation
- Includes a rotor that holds 36 x 1.5-2.0 ml tubes (RD36)
Operating Features:
- Consistent performance: designed to ensure consistent drying times and reproducible results
- User-friendly interface
- Three user defined programs that can be modified and saved
- Temperature set point: ambient, 35°C to 65°C in 5°C increments
- Heat Time setting: from 1 m to 9h 59 m or continuous
- Run Time setting: from 1m to 9h 59m
- Audible alarm to alert user when run is complete
Sample Protection
- Dry at ambient temperatures or using low heat to avoid sample denaturing
- Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no solvent bumping
Safety Features
- PTFE-coated chamber resists chemicals and cleans easily
- Latched lid and lid lock indicator ensures user safety during operation
- Lid stay mechanism to secure lid while it‘s open, allowing user to have both hands free for operation
- Unit and pump are virtually maintenance-free
- External post trap to neutralize ammonium hydroxide vapors
- Optional chemical traps and optional cartridges to trap radioactive chemicals to minimize concerns in handling hazaRDO™us fumes and chemicals.
Solvents and Solvent Combinations:
- Water
- PCR buffers (aqueous)
- Ethanol and water
- Ammonium Hydroxide (will require an ammonia post-trap ANT-100 and neutralizing solution accessory ANS121)
Specifications
UL Listed to US and Canadian requirements. CE marked | |
RD36 rotor (36x 1.5-2.0 ml tubes) | |
105 lb. | |
Ambient to 65°C | |
SpeedVac DNA130 Vacuum Concentrator System | |
39 kg |
Accommodates 36 x 1.5 mL Tubes, 72 x 0.5 mL Tubes and 2 Microwell Plates | |
<10 Torr (13 mbar, 1.3k Pa) | |
48 kg | |
PTFE-coated Aluminum | |
86 lb. | |
11.5 x 25 x 12 in. (29 x 64 x 31 cm) |