Learn More
Implen™ NanoPhotometer™ N60 Micro-Volume UV-VIS Spectrophotometer
The Implen NanoPhotometer™ N60 UV/Vis spectrophotometer for NanoVolume applications. Quantify DNA, RNA, proteins and scan other samples within seconds.
£8949.07 - £9465.36
Specifications
Absorbance Accuracy | < 1.75% at 0.7 A at 280 nm of the reading |
---|---|
Absorbance Range | 0.02 to 330 A |
Connections | 2x USB A, USB B, HDMI, Ethernet, WiFi |
Detector Type | 3648-element CCD Array |
Display | 7 in. Glove Compatible Touchscreen, 1024 x 600 pixels |
Product Code | Brand | Includes | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Model | Price | Quantity & Availability | ||||
15442203
|
Implen™
N60-TOUCH |
Power Supply, USB Cable, Dust Cover, Software & Manual | N60-Touch |
£8949.07
Each |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15715815
|
Implen™
N60-MOBILE |
Battery, Power Supply, USB Cable, Dust Cover, Software & Manual | N60-Mobile |
£9465.36
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Implen NanoPhotometer™ N60 UV/VIS spectrophotometer is the right instrument when it comes to the quantification of nucleic acids, proteins or cell cultures as it offers full scan data from 200 - 900 nm in just 0.3 - 2 μL of sample. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.
Disturbing surface tension and evaporation effects are eliminated due to the compression of the sample. No recalibration is necessary over the entire lifetime. Optional CFR21 software available for 21 CFR part 11 compliance for GxP environment.
- Detection range dsDNA 1 ng/μL - 16,500 ng/μL
- Detection range BSA 0.03 mg/mL - 478 mg/mL
- Minimum sample Size 0.3 μL
- Scan range 200 - 900 nm
- Integrated Vortex for tubes up to 2.0 mL
- Measure time for full scan range: 2.5 - 4.0 seconds
- Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA, Lowry), kinetics, OD600 and manual methods
- Auto Sample function starts automatically once sample arm is closed
- Recalibration-free due to True Path Technology™
- Independent from surface tension of sample due to Sample Compression Technology™
- Sample ControlTM and Blank ControlTM for analysis of sample quality and identification of contaminants
- Control options: Onboard with built-in 7 in. Touchscreen, Computer and Tablet
- In & Output Ports 2x USB A, USB B, HDMI, Ethernet, Wi-Fi
- 21 CFR part 11 compliant software (optional)
- Built-in file server
- LIMS integration through REST API
- Made in Germany
Specifications
< 1.75% at 0.7 A at 280 nm of the reading | |
2x USB A, USB B, HDMI, Ethernet, WiFi | |
7 in. Glove Compatible Touchscreen, 1024 x 600 pixels | |
English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese | |
<1.8 nm | |
200 to 900 nm | |
1 to 16,500 ng/μL | |
1 | |
Linux based NPOS | |
< 0.5% @ 240 nm using NaI | |
± 0.75 nm |
0.02 to 330 A | |
3648-element CCD Array | |
20 x 20 x 12 cm | |
Xenon Flash | |
90/250 V | |
Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom | |
2.5 to 4.0 sec | |
0.67 and 0.07 mm | |
0.3 to 2 μL | |
Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS |
For Research Use Only