missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 20 Blue

Catalog Number p-8349003
185.59 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.


Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to protect workers from certain substances with a high level of comfort. Dupont™ ProShield™ 20 Blue Hooded Coverall is typically used for protection against particles (Type 5) and limited liquid splashes or sprays (Type 6).

  • Chemical protective clothing, Category III, Type 5 and 6
  • EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched external seams
  • Nylon zipper with flap
  • Available in blue in sizes S to XXXL (a white garment is also available).
  • 2-piece hood.
  • Elasticated face, wrists, waist and ankles
TRUSTED_SUSTAINABILITY
Product Type Hooded Coveralls
Size Small
Material SMS Fabric
Color Blue
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper with Storm Flap
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-5
Size (Metric) Chest 84 to 92 cm
For Use With (Application) For workers seeking protection against dirt and grime during light duty work and other industries
Quantity 50/Pk.
Show More Show Less

The user should make the risk assessment to determine the protection required.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'