Learn More
p21 Mouse anti-Mouse, Unconjugated, Clone: 3C7D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67362-1-IG-150UL
This item is not returnable.
View return policy
Description
The p21 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of mouse p21. This antibody recognizes mouse antigen. The p21 antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
p21 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Mouse |
Immunofluorescence | |
3C7D3 | |
Unconjugated | |
150 μL | |
12575 | |
IgG2b |