missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA124622
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-24622 is expected to cross react with human (100% conserved) and mouse (100% conserved) due to sequence homology.
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with relatively high calcium permeability. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle, being responsible, for example, for sympathetic vasoconstriction in small arteries, arterioles and vas deferens. Mouse studies suggest that this receptor is essential for normal male reproductive function. It is possible that the development of selective antagonists for this receptor may provide an effective non-hormonal male contraceptive pill.Specifications
P2X1 | |
Polyclonal | |
Unconjugated | |
P2RX1 | |
AI323649; ATP receptor; BB122383; P2RX1; P2x; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; P2XMR; Pdcd3; programmed cell death 3; Purinergic receptor; purinergic receptor P2X 1; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; purinergic receptor P2X1-1; purinergic receptor P2X1-2; RP-2; RP-2 protein | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Frozen) | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P47824, P51575, P51576 | |
P2RX1 | |
Synthetic peptide corresponding to residues G(364) L Q E N M R T S(372) of rat P2X1. | |
RUO | |
18436, 25505, 5023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |