missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA120758
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-20758 detects P2Y12 in human samples.
P2Y12 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is involved in platelets aggregation, and is a potential target for the treatment of thromboembolisms and other clotting disorders. Two transcript variants encoding the same isoform have been identified for this gene. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries.Specifications
P2Y12 | |
Polyclonal | |
Unconjugated | |
P2ry12 | |
2900079B22Rik; 4921504D23Rik; ADP-glucose receptor; ADPG-R; BDPLT8; Gi-coupled ADP receptor HORK3; G-protein coupled receptor SP1999; HORK3; P2RY12; P2T(AC); P2Y purinoceptor 12; P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2y12; P2Y12 platelet ADP receptor; P2YR12; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled 12; purinergic receptor P2Y, G-protein coupled, 12; purinergic receptor P2Y12; putative G-protein coupled receptor; SP1999 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64805 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H244 | |
P2ry12 | |
Synthetic peptide, conjugated to KLH, corresponding to the N-terminal extracellular domain of human P2Y12. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |